This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247644_m1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247644_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 13 - 14 1635 72
NM_001037878.3 NP_001032967.1 13 - 14 1635 72
NM_001286362.1 NP_001273291.1 12 - 13 1560 72
NM_001286363.1 NP_001273292.1 12 - 13 1560 72
NM_025804.3 NP_080080.2 13 - 14 1635 72
GenBank mRNA AK003180.1 - 13 - 14 1522 72
AK013754.1 - 13 - 14 1324 72
AK045397.1 - 12 - 13 1488 72
AK050274.1 - 13 - 14 1561 72
AK076151.1 - 7 - 8 722 72
AK088360.1 - 13 - 14 1523 72
AK089964.1 - 13 - 14 1524 72
AK122430.1 - 13 - 14 1490 72
AK148082.1 - 13 - 14 1497 72
AK150918.1 - 13 - 14 1516 72
AK150942.1 - 13 - 14 1509 72
AK151143.1 - 13 - 14 1509 72
AK151558.1 - 13 - 14 1515 72
AK152178.1 - 13 - 14 1509 72
AK152494.1 - 13 - 14 1509 72
AK161627.1 - 13 - 14 1511 72
AK167837.1 - 13 - 14 1509 72
AK168167.1 - 13 - 14 1509 72
BC016130.1 - 13 - 14 1497 72
BC025071.1 - 13 - 14 1496 3498003 72
BC046768.1 - 13 - 14 1593 5707312 72
EU624323.1 - 12 - 13 1398 72
U94988.1 - 13 - 14 1496 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001520.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012464.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012464.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012464.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012464.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012464.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012464.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0