This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247648_m1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247648_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 3 - 4 591 115
NM_001037878.3 NP_001032967.1 3 - 4 591 115
NM_025804.3 NP_080080.2 3 - 4 591 115
GenBank mRNA AK003180.1 - 3 - 4 479 115
AK013754.1 - 3 - 4 280 115
AK050274.1 - 3 - 4 519 115
AK075728.1 - 3 - 4 445 115
AK088360.1 - 3 - 4 479 115
AK089964.1 - 3 - 4 480 115
AK148082.1 - 3 - 4 453 115
AK150232.1 - 3 - 4 478 115
AK151558.1 - 3 - 4 472 115
AK161627.1 - 3 - 4 467 115
AK167837.1 - 3 - 4 465 115
AK168167.1 - 3 - 4 465 115
BC025071.1 - 3 - 4 452 3498003 115
BC046768.1 - 3 - 4 549 5707312 115
U94988.1 - 3 - 4 115

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK013754.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012448.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012448.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012448.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012448.mm.1 TC0800001520.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800012449.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012449.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012449.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012449.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012449.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012449.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0