This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247721_m1
See other TTPAL GE Assays ›
Gene Symbol:
Ttpal
Gene Name:
tocopherol (alpha) transfer protein-like
Gene Aliases:
3110080A02Rik, 5830472M02Rik
Chromosome Location:
Chr.2: 163602281 - 163619013 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247721_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttpal
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029512.2 NP_083788.2 3 - 4 762 140
NM_181734.3 NP_859423.2 3 - 4 759 140
GenBank mRNA AK033600.1 - 3 - 4 708 140
AK039450.1 - 3 - 4 760 140
AK043791.1 - 3 - 4 729 140
AK081680.1 - 3 - 4 722 140
AK156641.1 - 3 - 4 725 140
BC052389.1 - 3 - 4 718 5718874 140
BC060726.1 - 2 - 3 628 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002491.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ttpal

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function
transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200020331.mm.1 NM_029512.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020331.mm.1 NM_029512.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200020331.mm.1 NM_181734.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020331.mm.1 NM_181734.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200020331.mm.1 TC0200002491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200020333.mm.1 NM_029512.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020333.mm.1 NM_029512.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200020333.mm.1 NM_181734.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020333.mm.1 NM_181734.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200020333.mm.1 TC0200002491.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0