This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247834_m1
See other SOCS2 GE Assays ›
Gene Symbol:
Socs2
Gene Name:
suppressor of cytokine signaling 2
Gene Aliases:
8030460M17, AI527257, AW108012, CIS2, Cish2, D130043N08Rik, JAB, SOCS-2, SSI-2, hg
Chromosome Location:
Chr.10: 95383903 - 95417253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247834_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Socs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168655.1 NP_001162126.1 1 - 2 212 139

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001168655.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Apoptosis Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
negative regulation of protein kinase activity
lactation
negative regulation of signal transduction
cytokine-mediated signaling pathway
response to estradiol
cellular response to hormone stimulus
intracellular signal transduction
regulation of growth
regulation of multicellular organism growth
negative regulation of multicellular organism growth
positive regulation of neuron differentiation
negative regulation of JAK-STAT cascade
negative regulation of insulin receptor signaling pathway
growth hormone receptor signaling pathway
mammary gland alveolus development
protein kinase inhibitor activity
growth hormone receptor binding
insulin-like growth factor receptor binding
JAK pathway signal transduction adaptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000020946.mm.1 NM_001168655.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020946.mm.1 NM_001168655.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000020946.mm.1 TC1000002725.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline