This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247991_m1
See other STAU1 GE Assays ›
Gene Symbol:
Stau1
Gene Name:
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases:
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location:
Chr.2: 166947549 - 166996299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247991_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stau1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109905.2 NP_001103375.1 3 - 4 569 118
NM_001109906.2 NP_001103376.1 3 - 4 569 118
NM_001291160.1 NP_001278089.1 3 - 4 569 118
NM_011490.4 NP_035620.1 3 - 4 569 118
NR_111895.1 2 - 3 303 118
GenBank mRNA AF061942.1 - 3 - 4 569 118
AF395842.1 - 2 - 3 290 118
AK005002.2 - 3 - 4 510 118
AK139532.1 - 3 - 4 516 118
AK145826.1 - 3 - 4 536 118
AK151835.1 - 3 - 4 375 118
AK152269.1 - 3 - 4 118
AK152744.1 - 3 - 4 375 118
AK159602.1 - 3 - 4 486 118
AK159781.1 - 3 - 4 539 118
AK160979.1 - 3 - 4 510 118
AK167637.1 - 3 - 4 510 118
AK193147.1 - 176 118
AY391773.1 - 2 - 3 302 118
BC012959.1 - 3 - 4 533 4188394 118
BC082277.1 - 3 - 4 506 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005179.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stau1

Gene Ontology Categories:

Function(s) Process(es)

intracellular mRNA localization
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
RNA binding
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200044312.mm.1 NM_001109905.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044312.mm.1 NM_001109906.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044312.mm.1 NM_011490.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200044312.mm.1 TC0200005179.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0