Assay Details
| Gene Symbol: Rbfox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024931.2 | NP_001020102.2 | 10 - 11 | 1214 | 136 | |
| NM_001039167.1 | NP_001034256.1 | 10 - 11 | 1214 | 136 | ||
| NM_001039168.1 | NP_001034257.1 | 10 - 11 | 1211 | 136 | ||
| NM_001285436.1 | NP_001272365.1 | 9 - 10 | 1174 | 136 | ||
| NM_001285437.1 | NP_001272366.1 | 9 - 10 | 1014 | 136 | ||
| NM_001285438.1 | NP_001272367.1 | 9 - 10 | 1020 | 136 | ||
| GenBank mRNA | AF229056.1 | - | 6 - 7 | 459 | 136 | |
| AK032676.1 | - | 9 - 10 | 1021 | 136 | ||
| AK049926.1 | - | 10 - 11 | 1193 | 136 | ||
| AK079086.1 | - | 10 - 11 | 1215 | 136 | ||
| AK086669.1 | - | 10 - 11 | 1195 | 138 | ||
| AK160816.1 | - | 9 - 10 | 1153 | 136 | ||
| BC054403.1 | - | 9 - 10 | 1014 | 6494361 | 136 | |
| FJ958310.1 | - | 6 - 7 | 628 | 136 | ||
| FJ958311.1 | - | 6 - 7 | 628 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbfox3
Genomic Map