Assay Details
| Gene Symbol: Rbfox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039167.1 | NP_001034256.1 | 11 - 12 | 1344 | 99 | |
| NM_001285436.1 | NP_001272365.1 | 10 - 11 | 1304 | 99 | ||
| NM_001285437.1 | NP_001272366.1 | 10 - 11 | 1144 | 99 | ||
| GenBank mRNA | AF229056.1 | - | 7 - 8 | 589 | 99 | |
| AK079086.1 | - | 11 - 12 | 1345 | 99 | ||
| AK160816.1 | - | 10 - 11 | 1283 | 99 | ||
| BC054403.1 | - | 10 - 11 | 1144 | 6494361 | 99 | |
| FJ958310.1 | - | 7 - 8 | 758 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbfox3
Genomic Map