This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01248772_m1
See other RBFOX3 GE Assays ›
Gene Symbol:
Rbfox3
Gene Name:
RNA binding protein, fox-1 homolog (C. elegans) 3
Gene Aliases:
Fox-3, Hrnbp3, NeuN, Neuna60
Chromosome Location:
Chr.11: 118489760 - 118911597 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01248772_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbfox3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039167.1 NP_001034256.1 14 - 15 1672 130
NM_001039168.1 NP_001034257.1 14 - 15 1528 130
GenBank mRNA AK079086.1 - 14 - 15 1673 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004164.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
RNA splicing
regulation of RNA splicing
nucleotide binding
nucleic acid binding
DNA binding
RNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0