Assay Details
| Gene Symbol: Rbfox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024931.2 | NP_001020102.2 | 1 - 2 | 276 | 119 | |
| NM_001039167.1 | NP_001034256.1 | 1 - 2 | 276 | 119 | ||
| NM_001039168.1 | NP_001034257.1 | 1 - 2 | 276 | 119 | ||
| NM_001285436.1 | NP_001272365.1 | 1 - 2 | 276 | 119 | ||
| GenBank mRNA | AK049926.1 | - | 1 - 2 | 254 | 119 | |
| AK079086.1 | - | 1 - 2 | 277 | 119 | ||
| AK086669.1 | - | 1 - 2 | 256 | 119 | ||
| AK160816.1 | - | 1 - 2 | 255 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK079086.1, AK160816.1, NM_001024931.2, NM_001039167.1, NM_001039168.1, NM_001285436.1 transcripts.
Set Membership: |
|

Rbfox3
Genomic Map