This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01248794_m1
See other MARCH10 GE Assays ›
Gene Symbol:
March10
Gene Name:
membrane-associated ring finger (C3HC4) 10
Gene Aliases:
4933417C16Rik, AV039223, OTTMUSG00000002847, Rnf190
Chromosome Location:
Chr.11: 105360798 - 105456863 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01248794_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: March10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039242.2 NP_001034331.1 4 - 5 563 73
NM_172568.2 NP_766156.2 4 - 5 563 73
GenBank mRNA AK132734.1 - 4 - 5 552 73
BC100474.1 - 4 - 5 556 30918723 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003907.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene March10

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100036490.mm.1 NM_172568.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036490.mm.1 NM_172568.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036490.mm.1 TC1100003907.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100036495.mm.1 NM_001039242.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036495.mm.1 NM_001039242.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036495.mm.1 NM_172568.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036495.mm.1 NM_172568.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036495.mm.1 TC1100003907.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100036488.mm.1 NM_172568.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036488.mm.1 NM_172568.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036488.mm.1 TC1100003907.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0