This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01248896_g1
See other GPT2 GE Assays ›
Gene Symbol:
Gpt2
Gene Name:
glutamic pyruvate transaminase (alanine aminotransferase) 2
Gene Aliases:
4631422C05Rik, ALT2, AU014768, AU041193, C87201
Chromosome Location:
Chr.8: 85492474 - 85527560 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01248896_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173866.3 NP_776291.1 1 - 2 75 154
GenBank mRNA AK031958.1 - 1 - 2 87 154
AK033424.1 - 1 - 2 76 154
AK075894.1 - 1 - 2 94 154
AK076250.1 - 1 - 2 93 154
AK082030.1 - 1 - 2 118 154
BC034219.1 - 1 - 2 107 5065322 154

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK033424.1, AK075894.1, AK076250.1, AK082030.1, BC034219.1, NM_173866.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpt2

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

2-oxoglutarate metabolic process
biosynthetic process
L-alanine metabolic process
catalytic activity
L-alanine:2-oxoglutarate aminotransferase activity
transaminase activity
transferase activity
pyridoxal phosphate binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 752946 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0