This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249323_g1
See other SPNS2 GE Assays ›
Gene Symbol:
Spns2
Gene Name:
spinster homolog 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 72451638 - 72489920 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249323_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spns2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276383.1 NP_001263312.1 12 - 13 1434 64
NM_153060.3 NP_694700.2 12 - 13 1782 64
GenBank mRNA AK156007.1 - 13 - 14 1428 64
BC011467.1 - 12 - 13 1782 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003146.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spns2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

B cell homeostasis
lymphocyte homeostasis
regulation of humoral immune response
sphingosine-1-phosphate signaling pathway
sphingolipid metabolic process
transport
lipid transport
T cell homeostasis
regulation of eye pigmentation
lymph node development
transmembrane transport
bone development
lymphocyte migration
sphingolipid transporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018929 BC025823.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018929 AK156007.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018929 BC011467.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018929 NM_153060.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018929 AK083908.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263754 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89696 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45908 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173946 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34501 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907204 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308816 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916088 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921391 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227964 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813723 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228453 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297247 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368224 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143906 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710633 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735447 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347156 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894941 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54522 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945152 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858928 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13752 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20931 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79282 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123565 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112375 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018929 BC025823.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018929 AK156007.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018929 BC011467.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018929 NM_153060.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018929 AK083908.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07393538 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07393589 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112375 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123565 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07393604 - -
Whole Mouse Genome G4122A A_51_P123745 BC025823.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P123745 AK156007.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P123745 BC011467.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P123745 NM_153060.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P123745 AK083908.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127141 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123565 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112375 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911717 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09605566 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911724 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911790 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911742 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911796 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911752 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911781 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911749 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911705 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09649083 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911793 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09637649 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911711 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09459043 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911728 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911739 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911710 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911661 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911693 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911726 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911771 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911664 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09649079 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911699 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911667 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09095805 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911645 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911731 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911798 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911684 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911802 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09095812 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09571008 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911761 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911785 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911642 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911801 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09461658 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911680 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911799 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911772 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911797 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09461642 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09724786 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09278011 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911671 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911806 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911783 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09492307 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911691 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911685 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09459038 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911727 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911720 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911725 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911688 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911763 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911753 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911813 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911737 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09397316 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911732 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911702 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09049423 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911805 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911778 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911715 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911769 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911676 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911740 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911748 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09461637 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911670 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09410681 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911729 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911733 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911779 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911788 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911795 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911775 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09134268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09361762 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911668 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911743 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911811 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911687 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09570998 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911745 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09361759 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09492300 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27911718 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10388199 - -
Affymetrix Mouse Gene 1.0 ST Array 10388210 - -
Affymetrix Mouse Gene 1.0 ST Array 10388203 - -
Affymetrix Mouse Gene 1.0 ST Array 10388209 - -
Affymetrix Mouse Gene 1.0 ST Array 10388196 - -
Affymetrix Mouse Gene 1.0 ST Array 10388208 - -
Affymetrix Mouse Gene 1.0 ST Array 10388206 - -
Affymetrix Mouse Gene 1.0 ST Array 10388207 - -
Affymetrix Mouse Gene 1.0 ST Array 10388200 - -
Affymetrix Mouse Gene 1.0 ST Array 10388198 - -
Affymetrix Mouse Gene 1.0 ST Array 10388195 - -
Affymetrix Mouse Gene 1.0 ST Array 10388197 - -
Affymetrix Mouse Gene 1.0 ST Array 10388205 - -
Affymetrix Mouse Gene 1.0 ST Array 10388202 - -
Affymetrix Mouse Gene 1.0 ST Array 10388204 - -
Affymetrix Mouse Gene 1.0 ST Array 10388201 - -
Affymetrix HT MG-430 PM Array Plate 1451601_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10388194 - -
Mouse Genome 430 2.0 Array 1451601_a_at - -
Mouse Genome 430 2.0 Array 1451601_PM_a_at - -
Affymetrix Mouse Clariom S Array PSR1100029175.mm.1 NM_001276383.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029175.mm.1 NM_153060.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100029175.mm.1 TC1100003146.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100029174.mm.1 NM_001276383.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029174.mm.1 NM_153060.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100029174.mm.1 TC1100003146.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100029173.mm.1 NM_001276383.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029173.mm.1 NM_153060.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100029173.mm.1 TC1100003146.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100029176.mm.1 NM_001276383.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029176.mm.1 NM_153060.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100029176.mm.1 TC1100003146.mm.1 MTA TC
Mouse Expression Set 430 1451601_PM_a_at - -
Mouse Expression Set 430 1451601_a_at - -
Mouse Genome 430A 2.0 Array 1451601_a_at - -
Mouse Genome 430A 2.0 Array 1451601_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1451601_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1451601_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 905012 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6620121 - -
Illumina mouse-6 v1.0 expression beadchip 6620121 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006620121 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660293 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2660293 - -
mouse-6 v1.1 (Illumina) scl39968.13_19-S - -
Illumina Sentrix mouse-6.v1 bead array 6620121 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl39968.13_19-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620121 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2660293 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0