This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249327_g1
See other SPNS2 GE Assays ›
Gene Symbol:
Spns2
Gene Name:
spinster homolog 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 72451638 - 72489920 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249327_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spns2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276383.1 NP_001263312.1 4 - 5 506 86
NM_153060.3 NP_694700.2 4 - 5 854 86
GenBank mRNA AB441166.1 - 4 - 5 726 86
AK156007.1 - 5 - 6 500 86
BC011467.1 - 4 - 5 854 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003146.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spns2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

B cell homeostasis
lymphocyte homeostasis
regulation of humoral immune response
sphingosine-1-phosphate signaling pathway
sphingolipid metabolic process
transport
lipid transport
T cell homeostasis
regulation of eye pigmentation
lymph node development
transmembrane transport
bone development
lymphocyte migration
sphingolipid transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100029191.mm.1 NM_001276383.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029191.mm.1 NM_153060.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100029191.mm.1 TC1100003146.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline