This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249387_g1
See other MRPL12 GE Assays ›
Gene Symbol:
Mrpl12
Gene Name:
mitochondrial ribosomal protein L12
Gene Aliases:
0610034O11Rik, 1500031N16Rik, MRP-L12, Rpml12
Chromosome Location:
Chr.11: 120484669 - 120488754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249387_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027204.2 NP_081480.2 1 - 2 98 94
GenBank mRNA AK002757.1 - 1 - 2 108 94
AK005328.1 - 1 - 2 129 95
AK028168.1 - 1 - 2 109 94
AK134816.1 - 1 - 2 131 94
AK166969.1 - 1 - 2 127 94
AK168337.1 - 1 - 2 101 94
BC005712.1 - 1 - 2 98 3582724 94
BC019565.1 - 1 - 2 96 4219731 94

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrpl12

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

transcription from mitochondrial promoter
translation
positive regulation of transcription, DNA-templated
structural constituent of ribosome

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 630492 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline