This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249598_g1
See other OGFOD1 GE Assays ›
Gene Symbol:
Ogfod1
Gene Name:
2-oxoglutarate and iron-dependent oxygenase domain containing 1
Gene Aliases:
4930415J21Rik, AA387199, AA939912, AW061076, mKIAA1612
Chromosome Location:
Chr.8: 94037141 - 94067923 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249598_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogfod1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177767.4 NP_808435.3 6 - 7 750 102
GenBank mRNA AK029629.1 - 6 - 7 752 102
AK122532.1 - 6 - 7 654 102
AK156765.1 - 6 - 7 720 102
AK166343.1 - 6 - 7 673 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001089.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogfod1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

regulation of translational termination
cell proliferation
protein hydroxylation
peptidyl-proline hydroxylation
stress granule assembly
oxidation-reduction process
sulfonate dioxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
L-ascorbic acid binding
peptidyl-proline dioxygenase activity
peptidyl-proline 3-dioxygenase activity
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800008398.mm.1 NM_177767.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008398.mm.1 NM_177767.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800008398.mm.1 TC0800001089.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0