This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250265_m1
See other TRAPPC2L GE Assays ›
Gene Symbol:
Trappc2l
Gene Name:
trafficking protein particle complex 2-like
Gene Aliases:
1810017G16Rik, AB030200, Hspc176, Tca17
Chromosome Location:
Chr.8: 122611626 - 122615591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250265_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc2l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021502.2 NP_067477.1 1 - 2 68 56
GenBank mRNA AB030200.1 - 1 - 2 61 56
AK007526.1 - 1 - 2 38 56
BC028502.1 - 1 - 2 48 1366538 56
BC114968.1 - 36 56
BY091059.1 - 1 - 2 69 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001494.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc2l

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s)

transport
ER to Golgi vesicle-mediated transport
vesicle-mediated transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012181.mm.1 NM_021502.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012181.mm.1 NM_021502.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800012181.mm.1 TC0800001494.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800012180.mm.1 NM_021502.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012180.mm.1 NM_021502.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800012180.mm.1 TC0800001494.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0