This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250267_m1
See other TRAPPC2L GE Assays ›
Gene Symbol:
Trappc2l
Gene Name:
trafficking protein particle complex 2-like
Gene Aliases:
1810017G16Rik, AB030200, Hspc176, Tca17
Chromosome Location:
Chr.8: 122611626 - 122615591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250267_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc2l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021502.2 NP_067477.1 4 - 5 408 86
GenBank mRNA AB030200.1 - 4 - 5 401 86
AK007526.1 - 4 - 5 378 86
BC028502.1 - 4 - 5 388 1366538 86
BC114968.1 - 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc2l

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s)

transport
ER to Golgi vesicle-mediated transport
vesicle-mediated transport

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234573 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234574 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234572 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234571 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 AK007526.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 BC028502.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 NM_021502.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 AB030200.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 AK007526.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 BC028502.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 NM_021502.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443359 AB030200.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689703 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9598 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711574 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697071 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174147 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 AK007526.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 BC028502.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 NM_021502.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 AB030200.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 AK007526.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 BC028502.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 NM_021502.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443359 AB030200.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234580 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234581 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234595 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234583 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234587 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234594 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234604 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234589 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234611 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234609 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234600 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234612 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234571 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234592 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234601 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234603 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234570 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234586 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234598 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234574 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234597 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234602 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234590 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234575 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234606 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234572 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26234577 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174147 - -
Whole Mouse Genome G4122A A_51_P443359 AK007526.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 BC028502.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 NM_021502.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 AB030200.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 AK007526.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 BC028502.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 NM_021502.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P443359 AB030200.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127649 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174147 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06672527 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06672516 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06672508 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06672507 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07330928 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234574 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234573 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234572 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26234571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234598 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26234658 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06838665 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234598 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234597 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06672504 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234589 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07333137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234577 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07088735 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234611 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234587 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234571 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234590 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234606 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234608 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07088747 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07088731 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234570 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234575 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07088729 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06838661 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07088739 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06672517 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07088742 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234574 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234581 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06672523 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26234572 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10576154 - -
Affymetrix Mouse Gene 1.0 ST Array 10576155 - -
Affymetrix Mouse Gene 1.0 ST Array 10576156 - -
Affymetrix Mouse Gene 1.0 ST Array 10576157 - -
Affymetrix Mouse Gene 1.0 ST Array 10576153 - -
Affymetrix HT MG-430 PM Array Plate 1417105_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10576152 - -
Mouse Genome 430 2.0 Array 1417105_PM_at - -
Mouse Genome 430 2.0 Array 1417105_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 59005_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 59005_at - -
Mouse Expression Set 430 1417105_at - -
Mouse Expression Set 430 1417105_PM_at - -
Murine Genome U74v2 Set 98626_at - -
Murine Genome U74v2 Set 98626_PM_at - -
Mouse Genome 430A 2.0 Array 1417105_PM_at - -
Mouse Genome 430A 2.0 Array 1417105_at - -
Affymetrix HT Mouse Genome 430A Array 1417105_at - -
Affymetrix HT Mouse Genome 430A Array 1417105_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 735363 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5420373 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005420373 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766315 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2766315 - -
mouse-6 v1.1 (Illumina) scl33220.5.1_30-S - -
Illumina Sentrix mouse-6.v1 bead array 5420373 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl33220.5.1_30-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420373 - -
MouseRef-8 v2 Expression BeadChip ILMN_2766315 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766315 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33220.5.1_30-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0