This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250273_m1
See other CDT1 GE Assays ›
Gene Symbol:
Cdt1
Gene Name:
chromatin licensing and DNA replication factor 1
Gene Aliases:
2610318F11Rik, AW545653, C76791, Ris2
Chromosome Location:
Chr.8: 122568015 - 122573130 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250273_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026014.3 NP_080290.3 6 - 7 1012 118
GenBank mRNA AB086655.1 - 6 - 7 991 118
AF477481.1 - 6 - 7 1012 118
AK012041.1 - 6 - 7 989 118
AK028287.1 - 6 - 7 984 118
AK049163.1 - 6 - 7 984 118
AK076527.1 - 5 - 6 1094 118
AK083015.1 - 6 - 7 987 118
BC013842.1 - 6 - 7 967 118
BC024485.1 - 3 - 4 263 118
BC048076.1 - 6 - 7 1012 6307476 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001493.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdt1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication checkpoint
DNA replication
cell cycle
regulation of DNA-dependent DNA replication initiation
regulation of nuclear cell cycle DNA replication
DNA binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012171.mm.1 NM_026014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012171.mm.1 NM_026014.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012171.mm.1 TC0800001493.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline