This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250313_m1
See other CDH15 GE Assays ›
Gene Symbol:
Cdh15
Gene Name:
cadherin 15
Gene Aliases:
AI323380, Cdh14, Mcad
Chromosome Location:
Chr.8: 122848374 - 122867397 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250313_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007662.2 NP_031688.2 2 - 3 312 78
GenBank mRNA BC157978.1 - 2 - 3 229 9007298 78
M74541.1 - 2 - 3 719 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh15

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline