This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250732_m1
See other RBL1 GE Assays ›
Gene Symbol:
Rbl1
Gene Name:
retinoblastoma-like 1 (p107)
Gene Aliases:
AW547426, PRB1, p107
Chromosome Location:
Chr.2: 157145893 - 157204534 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250732_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001139516.1 NP_001132988.1 8 - 9 1166 77
NM_011249.2 NP_035379.2 8 - 9 1166 77
GenBank mRNA AK147064.1 - 8 - 9 1167 77
AK156055.1 - 8 - 9 1145 77
BC060124.1 - 8 - 9 1133 6408844 77
U27177.1 - 8 - 9 1150 77
U27178.1 - 8 - 9 1150 77
U33320.1 - 8 - 9 1133 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005015.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbl1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
chromatin modification
regulation of lipid kinase activity
regulation of cell cycle
protein binding
transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042922.mm.1 NM_001139516.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042922.mm.1 NM_001139516.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042922.mm.1 NM_011249.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042922.mm.1 NM_011249.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042922.mm.1 TC0200005015.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0