This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01250872_g1
See other DLG4 GE Assays ›
Gene Symbol:
Dlg4
Gene Name:
discs, large homolog 4 (Drosophila)
Gene Aliases:
Dlgh4, PSD-95, PSD95, SAP90, SAP90A
Chromosome Location:
Chr.11: 70018455 - 70045532 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01250872_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dlg4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109752.1 NP_001103222.1 10 - 11 1495 75
NM_007864.3 NP_031890.1 10 - 11 1504 75
GenBank mRNA BC014807.1 - 10 - 11 1240 4501403 75
BQ959940.1 - 4 - 5 647 75
D50621.1 - 10 - 11 1246 75

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dlg4

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of receptor internalization
protein complex assembly
signal transduction
positive regulation of cytosolic calcium ion concentration
chemical synaptic transmission
nervous system development
locomotory behavior
synaptic vesicle maturation
social behavior
protein localization to synapse
locomotory exploration behavior
response to cocaine
establishment of protein localization
nucleotide phosphorylation
regulation of neuronal synaptic plasticity
regulation of long-term neuronal synaptic plasticity
positive regulation of synaptic transmission
neuromuscular process controlling balance
dendritic spine morphogenesis
vocalization behavior
AMPA glutamate receptor clustering
receptor localization to synapse
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
guanylate kinase activity
receptor binding
structural molecule activity
protein binding
protein C-terminus binding
kinase binding
protein kinase binding
protein phosphatase binding
PDZ domain binding
beta-1 adrenergic receptor binding
D1 dopamine receptor binding
P2Y1 nucleotide receptor binding
protein complex binding
acetylcholine receptor binding
glutamate receptor binding
ionotropic glutamate receptor binding
neurexin family protein binding
neuroligin family protein binding
scaffold protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100008157.mm.1 NM_001109752.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008157.mm.1 NM_001109752.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008157.mm.1 NM_007864.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008157.mm.1 NM_007864.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100008157.mm.1 TC1100000937.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0