This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251051_m1
See other SLC9A5 GE Assays ›
Gene Symbol:
Slc9a5
Gene Name:
solute carrier family 9 (sodium/hydrogen exchanger), member 5
Gene Aliases:
Gm696
Chromosome Location:
Chr.8: 105348104 - 105369881 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251051_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc9a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081332.1 NP_001074801.1 12 - 13 2971 55
GenBank mRNA BC151197.1 - 12 - 13 2412 9088085 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001215.mm.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
proton transport
sodium ion transmembrane transport
hydrogen ion transmembrane transport
sodium:proton antiporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038257 AK163918.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038257 NM_001081332.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596584 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570692 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853694 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738339 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881944 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219564 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53228 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721980 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389430 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878419 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92808 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4681 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33052 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143810 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56102 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038257 AK163918.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038257 NM_001081332.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01817632 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06626418 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06626385 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01817632 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07335335 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07335325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133311 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133307 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133317 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133228 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133255 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133365 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133235 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133271 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133254 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133372 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133228 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133286 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133314 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133313 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133259 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133373 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07335313 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133305 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133294 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133279 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133300 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07174033 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133289 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133246 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133297 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133243 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133240 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133255 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133215 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133238 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07222390 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07335318 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07109548 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133317 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133267 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133281 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133233 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133295 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07000416 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06817999 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133253 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133368 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07000412 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133242 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07174029 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133219 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07321533 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133357 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133355 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07222386 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133310 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133307 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133298 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06708680 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133277 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133261 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06945388 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07174028 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133284 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06708685 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133359 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06708695 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26133239 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10574728 - -
Affymetrix Mouse Gene 1.0 ST Array 10574733 - -
Affymetrix Mouse Gene 1.0 ST Array 10574729 - -
Affymetrix Mouse Gene 1.0 ST Array 10574739 - -
Affymetrix Mouse Gene 1.0 ST Array 10574738 - -
Affymetrix Mouse Gene 1.0 ST Array 10574741 - -
Affymetrix Mouse Gene 1.0 ST Array 10574732 - -
Affymetrix Mouse Gene 1.0 ST Array 10574735 - -
Affymetrix Mouse Gene 1.0 ST Array 10574737 - -
Affymetrix Mouse Gene 1.0 ST Array 10574736 - -
Affymetrix Mouse Gene 1.0 ST Array 10574731 - -
Affymetrix Mouse Gene 1.0 ST Array 10574734 - -
Affymetrix Mouse Gene 1.0 ST Array 10574742 - -
Affymetrix Mouse Gene 1.0 ST Array 10574740 - -
Affymetrix Mouse Gene 1.0 ST Array 10574730 - -
Affymetrix HT MG-430 PM Array Plate 1455760_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1420070_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10574727 - -
Mouse Genome 430 2.0 Array 1449722_PM_at - -
Mouse Genome 430 2.0 Array 1449722_at - -
Mouse Genome 430 2.0 Array 1420070_a_at - -
Mouse Genome 430 2.0 Array 1455760_at - -
Mouse Genome 430 2.0 Array 1455760_PM_at - -
Mouse Genome 430 2.0 Array 1420070_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 277973_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 277973_PM_at - -
Affymetrix Mouse Clariom S Array PSR0800009601.mm.1 NM_001081332.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009601.mm.1 NM_001081332.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009601.mm.1 TC0800001215.mm.1 MTA TC
Mouse Expression Set 430 1420070_PM_a_at - -
Mouse Expression Set 430 1420070_a_at - -
Mouse Expression Set 430 1455760_at - -
Mouse Expression Set 430 1455760_PM_at - -
Murine Genome U74v2 Set 140439_at - -
Murine Genome U74v2 Set 140439_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1455760_at - -
Affymetrix HT Mouse Genome 430B Array 1420070_a_at - -
Affymetrix HT Mouse Genome 430B Array 1455760_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1420070_PM_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 473288 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_1223654 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bm6jx:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0