This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251063_m1
See other SLC9A5 GE Assays ›
Gene Symbol:
Slc9a5
Gene Name:
solute carrier family 9 (sodium/hydrogen exchanger), member 5
Gene Aliases:
Gm696
Chromosome Location:
Chr.8: 105348104 - 105369881 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251063_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc9a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081332.1 NP_001074801.1 9 - 10 2621 67
GenBank mRNA BC151197.1 - 9 - 10 2062 9088085 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001215.mm.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
proton transport
sodium ion transmembrane transport
hydrogen ion transmembrane transport
sodium:proton antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800009598.mm.1 NM_001081332.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009598.mm.1 NM_001081332.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009598.mm.1 TC0800001215.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0