This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251249_m1
See other DUSP15 GE Assays ›
Gene Symbol:
Dusp15
Gene Name:
dual specificity phosphatase-like 15
Gene Aliases:
AI851682, LMW-DSP10, T-DSP10
Chromosome Location:
Chr.2: 152940995 - 152952277 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251249_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dusp15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159376.1 NP_001152848.1 5 - 6 486 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004920.mm.1

Set Membership:

Amplicon length between 71 and 85 Protein Phosphatase Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
transforming growth factor beta receptor signaling pathway
dephosphorylation
regulation of cell proliferation
positive regulation of JNK cascade
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200041709.mm.1 NM_001159376.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200041709.mm.1 NM_001159376.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200041709.mm.1 TC0200004920.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline