This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251347_m1
See other BCO1 GE Assays ›
Gene Symbol:
Bco1
Gene Name:
beta-carotene oxygenase 1
Gene Aliases:
Bcdo, Bcdo1, Bcmo1, CMO1, Cmoi, beta-CD, betaCMOOX
Chromosome Location:
Chr.8: 117095851 - 117133720 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251347_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bco1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163028.1 NP_001156500.1 2 - 3 362 69
NM_021486.3 NP_067461.2 2 - 3 362 69
GenBank mRNA AF294899.1 - 2 - 3 375 69
AJ278064.2 - 2 - 3 374 69
AK054171.1 - 1 - 2 114 69
AK146472.1 - 2 - 3 359 69
BC125328.1 - 2 - 3 229 40129843 69
BC125330.1 - 2 - 3 229 40129845 69
BC145036.1 - 2 - 3 229 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001396.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bco1

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
oxidation-reduction process
beta-carotene 15,15'-monooxygenase activity
monooxygenase activity
oxidoreductase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011330.mm.1 NM_001163028.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011330.mm.1 NM_001163028.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011330.mm.1 NM_021486.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011330.mm.1 NM_021486.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011330.mm.1 TC0800001396.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011331.mm.1 NM_001163028.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011331.mm.1 NM_001163028.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011331.mm.1 NM_021486.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011331.mm.1 NM_021486.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011331.mm.1 TC0800001396.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0