This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251435_m1
See other DLGAP4 GE Assays ›
Gene Symbol:
Dlgap4
Gene Name:
discs, large homolog-associated protein 4 (Drosophila)
Gene Aliases:
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16
Chromosome Location:
Chr.2: 156613633 - 156764363 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251435_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dlgap4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277186.1 NP_001264115.1 4 - 5 1661 62
NM_001277187.1 NP_001264116.1 4 - 5 1661 62
NM_146128.6 NP_666240.4 4 - 5 1661 62
GenBank mRNA AK122408.1 - 4 - 5 1661 62
AY243849.2 - 2 - 3 1241 62
BC145394.1 - 2 - 3 1298 9053952 62
BC145395.1 - 2 - 3 1298 9053953 62
CX210973.1 - 2 - 3 445 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002398.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dlgap4

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
signaling
protein binding
protein domain specific binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200019339.mm.1 NM_001277186.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200019339.mm.1 NM_001277187.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200019339.mm.1 NM_146128.6 Refseq
Affymetrix Mouse Clariom S Array PSR0200019339.mm.1 TC0200002398.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7txcj:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline