This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251702_m1
See other MRI1 GE Assays ›
Gene Symbol:
Mri1
Gene Name:
methylthioribose-1-phosphate isomerase homolog (S. cerevisiae)
Gene Aliases:
2410018C20Rik
Chromosome Location:
Chr.8: 84249629 - 84257337 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251702_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mri1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026423.4 NP_080699.3 2 - 3 605 89
GenBank mRNA AK007709.1 - 2 - 3 566 89
AK010538.1 - 2 - 3 577 89
AK013882.1 - 2 - 3 574 89
AK144807.1 - 2 - 3 546 89
AK164894.1 - 2 - 3 574 89
BC025049.1 - 2 - 3 572 4951045 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002579.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mri1

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid biosynthetic process
methionine biosynthetic process
L-methionine biosynthetic process from methylthioadenosine
cellular metabolic process
cellular biosynthetic process
isomerase activity
identical protein binding
S-methyl-5-thioribose-1-phosphate isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800019668.mm.1 NM_026423.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019668.mm.1 NM_026423.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800019668.mm.1 TC0800002579.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019666.mm.1 NM_026423.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019666.mm.1 NM_026423.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800019666.mm.1 TC0800002579.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline