This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251976_g1
See other P2RX1 GE Assays ›
Gene Symbol:
P2rx1
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 1
Gene Aliases:
AI323649, BB122383, P2x, Pdcd3, RP-2
Chromosome Location:
Chr.11: 72999056 - 73015200 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251976_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008771.3 NP_032797.3 9 - 10 1137 86
GenBank mRNA AK007650.1 - 3 - 4 504 86
AK035304.1 - 9 - 10 1138 86
BC015084.1 - 9 - 10 1151 4189541 86
X84896.1 - 9 - 10 1131 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2rx1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

serotonin secretion by platelet
regulation of vascular smooth muscle contraction
transport
ion transport
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of smooth muscle contraction
insemination
regulation of blood pressure
response to organic substance
neuronal action potential
regulation of vasoconstriction
platelet activation
response to ATP
synaptic transmission, glutamatergic
purinergic nucleotide receptor signaling pathway
vasoconstriction
positive regulation of ion transport
ceramide biosynthetic process
protein homooligomerization
protein heterooligomerization
regulation of calcium ion transport
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
cation channel activity
ATP binding
drug binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0