This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01252177_m1
See other FDXR GE Assays ›
Gene Symbol:
Fdxr
Gene Name:
ferredoxin reductase
Gene Aliases:
-
Chromosome Location:
Chr.11: 115268024 - 115277095 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01252177_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fdxr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007997.1 NP_032023.1 10 - 11 1262 66
GenBank mRNA AK134068.1 - 10 - 11 1253 66
BC057146.1 - 10 - 11 1196 66
BC071268.1 - 10 - 11 1262 6517167 66
D49920.1 - 10 - 11 1262 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004063.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fdxr

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
ubiquinone biosynthetic process
transport
steroid metabolic process
cholesterol metabolic process
oxidation-reduction process
NADPH oxidation
NADPH-adrenodoxin reductase activity
oxidoreductase activity
NADPH binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100038147.mm.1 NM_007997.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038147.mm.1 NM_007997.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038147.mm.1 TC1100004063.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0