This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01252565_m1
See other PRSS54 GE Assays ›
Gene Symbol:
Prss54
Gene Name:
protease, serine 54
Gene Aliases:
4931432M23Rik, Klkbl4
Chromosome Location:
Chr.8: 95559067 - 95576330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01252565_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss54
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027640.1 NP_081916.1 1 - 2 62 67
GenBank mRNA AK016501.1 - 3 - 4 413 67
BC144859.1 - 2 - 3 189 9053407 67
BC145787.1 - 2 - 3 189 40131090 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002747.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
biological_process
molecular_function
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021124.mm.1 TC0800002747.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021123.mm.1 NM_027640.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021123.mm.1 NM_027640.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021123.mm.1 TC0800002747.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021116.mm.1 TC0800002747.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021122.mm.1 NM_027640.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021122.mm.1 NM_027640.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021122.mm.1 TC0800002747.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021125.mm.1 TC0800002747.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline