This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253139_m1
See other SLC10A7 GE Assays ›
Gene Symbol:
Slc10a7
Gene Name:
solute carrier family 10 (sodium/bile acid cotransporter family), member 7
Gene Aliases:
2410193C02Rik, P7
Chromosome Location:
Chr.8: 78509328 - 78734012 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253139_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc10a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009981.2 NP_001009981.1 9 - 10 1010 64
NM_001282109.1 NP_001269038.1 9 - 10 1018 64
NM_029736.2 NP_084012.1 10 - 11 1094 64
GenBank mRNA AK010834.1 - 10 - 11 1044 64
AK040517.1 - 10 - 11 973 64
AK152752.1 - 10 - 11 1055 64
AK152849.1 - 10 - 11 1055 64
AK165983.1 - 9 - 10 955 64
AY825926.1 - 10 - 11 847 64
AY825928.1 - 9 - 10 763 64
BC116666.1 - 10 - 11 960 40107077 64
BC116717.1 - 10 - 11 960 40107078 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc10a7

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
biological_process
molecular_function
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0