This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253142_m1
See other SLC10A7 GE Assays ›
Gene Symbol:
Slc10a7
Gene Name:
solute carrier family 10 (sodium/bile acid cotransporter family), member 7
Gene Aliases:
2410193C02Rik, P7
Chromosome Location:
Chr.8: 78509328 - 78734012 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253142_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc10a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009981.2 NP_001009981.1 2 - 3 427 108
NM_001282108.1 NP_001269037.1 2 - 3 427 108
NM_001282109.1 NP_001269038.1 2 - 3 427 108
NM_029736.2 NP_084012.1 2 - 3 427 108
GenBank mRNA AK010834.1 - 2 - 3 377 108
AK040517.1 - 2 - 3 306 108
AK079901.1 - 2 - 3 404 108
AK152752.1 - 2 - 3 388 108
AK152849.1 - 2 - 3 388 108
AK165983.1 - 2 - 3 364 108
AY825926.1 - 2 - 3 180 108
AY825928.1 - 2 - 3 180 108
BC116666.1 - 2 - 3 293 40107077 108
BC116717.1 - 2 - 3 293 40107078 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001282109.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc10a7

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
biological_process
molecular_function
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline