This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253359_g1
See other PYCR1 GE Assays ›
Gene Symbol:
Pycr1
Gene Name:
pyrroline-5-carboxylate reductase 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 120635712 - 120643782 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253359_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pycr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144795.3 NP_659044.1 5 - 6 1079 108
GenBank mRNA AK139185.1 - 5 - 6 1080 108
BC006727.1 - 6 - 7 724 3962093 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004222.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pycr1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

proline biosynthetic process
cellular amino acid biosynthetic process
cellular response to oxidative stress
regulation of mitochondrial membrane potential
oxidation-reduction process
negative regulation of hydrogen peroxide-induced cell death
pyrroline-5-carboxylate reductase activity
oxidoreductase activity
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100040377.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040377.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040377.mm.1 TC1100004222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100040376.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040376.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040376.mm.1 TC1100004222.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0