This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253489_m1
See other ARV1 GE Assays ›
Gene Symbol:
Arv1
Gene Name:
ARV1 homolog (yeast)
Gene Aliases:
1110067L22Rik, AI461928, AW121084
Chromosome Location:
Chr.8: 124722138 - 124740648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253489_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arv1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026855.4 NP_081131.1 2 - 3 330 77
GenBank mRNA AK004398.1 - 2 - 3 331 77
BC119376.1 - 2 - 3 310 8734125 77
BC120631.1 - 2 - 3 310 40129554 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001547.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arv1

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
bile acid metabolic process
sterol metabolic process
cholesterol transport
intracellular sterol transport
regulation of cholesterol metabolic process
regulation of plasma membrane sterol distribution
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012695.mm.1 NM_026855.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012695.mm.1 NM_026855.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800012695.mm.1 TC0800001547.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800012696.mm.1 NM_026855.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012696.mm.1 NM_026855.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800012696.mm.1 TC0800001547.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0