This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253587_m1
See other WWP2 GE Assays ›
Gene Symbol:
Wwp2
Gene Name:
WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases:
1300010O06Rik, AA690238, AIP2, AW554328
Chromosome Location:
Chr.8: 107436368 - 107558597 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253587_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Wwp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025830.3 NP_080106.1 17 - 18 1897 61
GenBank mRNA AK004962.2 - 17 - 18 1913 61
AK088936.1 - 17 - 18 1876 61
AK146306.1 - 16 - 17 1759 61
AK151548.1 - 17 - 18 1891 61
AK159248.1 - 8 - 9 941 61
AK183794.1 - 1 - 2 61 61
BC004712.1 - 2 - 3 217 61
BC018496.1 - 1 - 2 1095 61
BC039921.1 - 17 - 18 1889 4954925 61
BC048184.1 - 17 - 18 1897 6827381 61
HQ896246.1 - 8 - 9 994 61
JN712751.1 - 17 - 18 1930 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001274.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Wwp2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
negative regulation of gene expression
protein ubiquitination
negative regulation of transporter activity
regulation of ion transmembrane transport
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of protein transport
protein autoubiquitination
protein K63-linked ubiquitination
regulation of potassium ion transmembrane transporter activity
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription factor binding
ubiquitin-protein transferase activity
protein binding
transcription factor binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800010326.mm.1 NM_025830.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010326.mm.1 NM_025830.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800010326.mm.1 TC0800001274.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0