This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253766_g1
See other PNLIP GE Assays ›
Gene Symbol:
Pnlip
Gene Name:
pancreatic lipase
Gene Aliases:
1810007A24Rik, PL, PTL
Chromosome Location:
Chr.19: 58670365 - 58681788 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253766_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnlip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026925.3 NP_081201.2 4 - 5 354 72
GenBank mRNA AK007357.1 - 4 - 5 355 72
AY387690.1 - 3 - 4 326 72
BC061061.1 - 4 - 5 354 30316303 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000838.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnlip

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
post-embryonic development
lipid catabolic process
intestinal cholesterol absorption
positive regulation of triglyceride lipase activity
triglyceride lipase activity
lipase activity
hydrolase activity
metal ion binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007279.mm.1 NM_026925.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007279.mm.1 NM_026925.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007279.mm.1 TC1900000838.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007280.mm.1 NM_026925.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007280.mm.1 NM_026925.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007280.mm.1 TC1900000838.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0