Assay Details
| Gene Symbol: Smarcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018817.2 | NP_061287.2 | 4 - 5 | 1215 | 79 | |
| GenBank mRNA | AF088884.1 | - | 4 - 5 | 1195 | 79 | |
| AK035854.1 | - | 4 - 5 | 1192 | 79 | ||
| AK042332.1 | - | 4 - 5 | 1194 | 79 | ||
| AK077878.1 | - | 4 - 5 | 1193 | 79 | ||
| AK083488.1 | - | 4 - 5 | 1193 | 79 | ||
| AK146767.1 | - | 4 - 5 | 1209 | 79 | ||
| AK154020.1 | - | 4 - 5 | 1198 | 79 | ||
| AK161570.1 | - | 4 - 5 | 1135 | 79 | ||
| AK169465.1 | - | 4 - 5 | 1206 | 79 | ||
| BC029078.1 | - | 4 - 5 | 1150 | 3992325 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcal1
Genomic Map