This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253859_mH
See other PDE4B GE Assays ›
Gene Symbol:
Pde4b
Gene Name:
phosphodiesterase 4B, cAMP specific
Gene Aliases:
Dpde4, R74983, dunce
Chromosome Location:
Chr.4: 102087537 - 102607262 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253859_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177983.1 NP_001171454.1 1 - 2 712 57
NM_019840.2 NP_062814.2 1 - 2 515 57
GenBank mRNA AF208023.2 - 1 - 2 450 57
AF326555.1 - 1 - 2 255 57
AF326556.1 - 1 - 2 255 57
AJ297397.1 - 1 - 2 450 57
BC049864.1 - 1 - 2 713 57
CB519636.1 - 1 - 2 515 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001047.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde4b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

neutrophil homeostasis
cAMP catabolic process
smooth muscle contraction
neutrophil chemotaxis
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
cellular response to drug
T cell receptor signaling pathway
leukocyte migration
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
regulation of cardiac muscle cell contraction
regulation of high voltage-gated calcium channel activity
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cAMP binding
ion channel binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400007126.mm.1 NM_001177983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007126.mm.1 NM_001177983.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007126.mm.1 NM_019840.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007126.mm.1 NM_019840.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400007126.mm.1 TC0400001047.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400007120.mm.1 NM_001177983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007120.mm.1 NM_001177983.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007120.mm.1 NM_019840.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007120.mm.1 NM_019840.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400007120.mm.1 TC0400001047.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400007118.mm.1 NM_001177983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007118.mm.1 NM_001177983.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007118.mm.1 TC0400001047.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400007121.mm.1 NM_001177983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007121.mm.1 NM_001177983.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007121.mm.1 NM_019840.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007121.mm.1 NM_019840.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400007121.mm.1 TC0400001047.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400007119.mm.1 NM_001177983.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007119.mm.1 NM_001177983.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007119.mm.1 NM_019840.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007119.mm.1 NM_019840.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400007119.mm.1 TC0400001047.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline