This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254017_g1
See other SLC7A9 GE Assays ›
Gene Symbol:
Slc7a9
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 9
Gene Aliases:
CSNU3
Chromosome Location:
Chr.7: 35448728 - 35466034 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254017_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199015.1 NP_001185944.1 6 - 7 812 94
NM_001199016.1 NP_001185945.1 5 - 6 700 94
NM_021291.3 NP_067266.1 5 - 6 927 94
GenBank mRNA AF192310.1 - 6 - 7 812 94
AJ249198.1 - 5 - 6 927 94
AK142541.1 - 5 - 6 701 94
AK164714.1 - 4 - 5 1117 94
BC010746.1 - 6 - 7 790 4236578 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000661.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a9

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
neutral amino acid transport
L-cystine transport
L-alpha-amino acid transmembrane transport
protein binding
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-cystine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 TC0700000661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 NM_001199015.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 NM_001199015.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 NM_001199016.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 NM_001199016.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 NM_021291.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005601.mm.1 NM_021291.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 TC0700000661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 NM_001199015.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 NM_001199015.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 NM_001199016.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 NM_001199016.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 NM_021291.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005599.mm.1 NM_021291.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 TC0700000661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 NM_001199015.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 NM_001199015.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 NM_001199016.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 NM_001199016.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 NM_021291.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700005598.mm.1 NM_021291.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0