This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254018_m1
See other SLC7A9 GE Assays ›
Gene Symbol:
Slc7a9
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 9
Gene Aliases:
CSNU3
Chromosome Location:
Chr.7: 35448728 - 35466034 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254018_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199015.1 NP_001185944.1 7 - 8 913 109
NM_001199016.1 NP_001185945.1 6 - 7 801 109
NM_021291.3 NP_067266.1 6 - 7 1028 109
GenBank mRNA AF192310.1 - 7 - 8 913 109
AJ249198.1 - 6 - 7 1028 109
AK142541.1 - 6 - 7 802 109
AK164714.1 - 5 - 6 1218 109
BC010746.1 - 7 - 8 891 4236578 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a9

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
neutral amino acid transport
L-cystine transport
L-alpha-amino acid transmembrane transport
protein binding
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-cystine transmembrane transporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline