This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254324_m1
See other ABLIM1 GE Assays ›
Gene Symbol:
Ablim1
Gene Name:
actin-binding LIM protein 1
Gene Aliases:
2210411C18Rik, 2610209L21Rik, 4833406P10Rik, 9330196J19Rik, AV079770, AW060987, AW215784, Limab1, mKIAA0059
Chromosome Location:
Chr.19: 57033264 - 57239560 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254324_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ablim1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103177.2 NP_001096647.1 10 - 11 1367 66
NM_001103178.2 NP_001096648.1 7 - 8 789 66
NM_001290813.1 NP_001277742.1 10 - 11 1353 66
NM_001290815.1 NP_001277744.1 10 - 11 1367 66
NM_001290816.1 NP_001277745.1 10 - 11 1367 66
NM_178688.3 NP_848803.3 12 - 13 1485 66
GenBank mRNA AF404774.1 - 12 - 13 1485 66
AF404775.1 - 10 - 11 1127 66
AF404776.1 - 7 - 8 749 66
AK079331.1 - 6 - 7 607 66
AK122196.1 - 10 - 11 1367 66
AK143907.1 - 10 - 11 1137 66
AK171860.1 - 1264 66
BC032216.1 - 1 - 2 162 66
BC141083.1 - 10 - 11 1347 9055695 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC032216.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ablim1

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
axon guidance
lamellipodium assembly
cilium assembly
positive regulation of transcription from RNA polymerase II promoter
actin binding
protein binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900015148.mm.1 NM_001103177.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900015148.mm.1 NM_001103178.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900015148.mm.1 NM_178688.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900015148.mm.1 NM_178688.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900015148.mm.1 TC1900001711.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0