This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254371_g1
See other TMBIM1 GE Assays ›
Gene Symbol:
Tmbim1
Gene Name:
transmembrane BAX inhibitor motif containing 1
Gene Aliases:
2310061B02Rik, AA960455, AU024746, C78899, RECS1, Tmbib1, mKIAA4161
Chromosome Location:
Chr.1: 74288247 - 74304336 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254371_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmbim1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027154.5 NP_081430.3 9 - 10 890 82
GenBank mRNA AB097685.1 - 8 - 9 636 82
AK077689.1 - 9 - 10 715 82
AK144110.1 - 9 - 10 734 82
AK147297.1 - 8 - 9 9598 82
AK152864.1 - 9 - 10 733 82
AK166926.1 - 8 - 9 1132 82
AK167286.1 - 4 - 5 189 82
AK170625.1 - 9 - 10 700 82
AK171102.1 - 9 - 10 732 82
AK186797.1 - 4 - 5 246 82
AK188714.1 - 4 - 5 233 82
AK220296.1 - 7 - 8 1896 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002583.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmbim1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
negative regulation of establishment of protein localization to plasma membrane
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of Fas signaling pathway
positive regulation of blood vessel remodeling
death receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100020801.mm.1 NM_027154.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100020801.mm.1 NM_027154.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100020801.mm.1 TC0100002583.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100020800.mm.1 NM_027154.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100020800.mm.1 NM_027154.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100020800.mm.1 TC0100002583.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0