This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254379_g1
See other TMBIM1 GE Assays ›
Gene Symbol:
Tmbim1
Gene Name:
transmembrane BAX inhibitor motif containing 1
Gene Aliases:
2310061B02Rik, AA960455, AU024746, C78899, RECS1, Tmbib1, mKIAA4161
Chromosome Location:
Chr.1: 74288247 - 74304336 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254379_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmbim1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027154.5 NP_081430.3 5 - 6 667 68
GenBank mRNA AB097685.1 - 4 - 5 413 68
AK077689.1 - 5 - 6 492 68
AK144110.1 - 5 - 6 511 68
AK147297.1 - 4 - 5 9375 68
AK152864.1 - 5 - 6 510 68
AK166926.1 - 5 - 6 683 68
AK170625.1 - 5 - 6 477 68
AK171102.1 - 5 - 6 509 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002583.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmbim1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
negative regulation of establishment of protein localization to plasma membrane
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of Fas signaling pathway
positive regulation of blood vessel remodeling
death receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100020807.mm.1 NM_027154.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100020807.mm.1 NM_027154.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100020807.mm.1 TC0100002583.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0