This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01255082_m1
See other DERA GE Assays ›
Gene Symbol:
Dera
Gene Name:
2-deoxyribose-5-phosphate aldolase homolog (C. elegans)
Gene Aliases:
2010002D22Rik, 2500002K03Rik, DEOC
Chromosome Location:
Chr.6: 137754577 - 137837872 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01255082_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dera
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172733.1 NP_766321.1 4 - 5 408 58
GenBank mRNA AK012558.1 - 4 - 5 444 58
AK034347.1 - 410 58
AK075963.1 - 4 - 5 433 58
AK083298.1 - 4 - 5 408 58
AK145820.1 - 4 - 5 444 58
AK150273.1 - 4 - 5 434 58
AK150712.1 - 4 - 5 404 58
AK152462.1 - 4 - 5 412 58
AK169418.1 - 4 - 5 428 58
BC016218.1 - 4 - 5 414 4484163 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001637.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dera

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

deoxyribonucleotide catabolic process
carbohydrate catabolic process
deoxyribonucleoside catabolic process
catalytic activity
deoxyribose-phosphate aldolase activity
lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600013045.mm.1 TC0600001637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013045.mm.1 NM_172733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013045.mm.1 NM_172733.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0