This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01255487_m1
See other BOLL GE Assays ›
Gene Symbol:
Boll
Gene Name:
bol, boule-like (Drosophila)
Gene Aliases:
4930554P13Rik, 4930597B14Rik
Chromosome Location:
Chr.1: 55239385 - 55364058 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01255487_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Boll
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113367.1 NP_001106838.1 2 - 3 369 137
NM_029267.3 NP_083543.2 2 - 3 314 137
GenBank mRNA AF272859.1 - 2 - 3 177 137
AK016125.1 - 2 - 3 315 137
AK016403.1 - 2 - 3 369 137

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001113367.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Boll

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
meiotic nuclear division
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of translational initiation
nucleotide binding
nucleic acid binding
RNA binding
translation activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100019165.mm.1 NM_001113367.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019165.mm.1 NM_001113367.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019165.mm.1 NM_029267.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019165.mm.1 NM_029267.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100019165.mm.1 TC0100002357.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0