Assay Details
| Gene Symbol: Boll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113367.1 | NP_001106838.1 | 4 - 5 | 513 | 111 | |
| NM_029267.3 | NP_083543.2 | 4 - 5 | 458 | 111 | ||
| GenBank mRNA | AF272859.1 | - | 4 - 5 | 321 | 111 | |
| AK016125.1 | - | 4 - 5 | 459 | 111 | ||
| AK016403.1 | - | 4 - 5 | 513 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113367.1 transcripts.
Set Membership: |
|

Boll
Genomic Map