This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01256065_g1
See other PLAA GE Assays ›
Gene Symbol:
Plaa
Gene Name:
phospholipase A2, activating protein
Gene Aliases:
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3
Chromosome Location:
Chr.4: 94565139 - 94603247 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01256065_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172695.2 NP_766283.2 3 - 4 667 90
GenBank mRNA AK019070.1 - 3 - 4 672 90
AK075934.1 - 3 - 4 599 90
AK162534.1 - 2 - 3 2858 90
AK166122.1 - 3 - 4 572 90
BC139355.1 - 3 - 4 666 8862377 90
BC139356.1 - 3 - 4 666 8862378 90
BC139773.1 - 3 - 4 577 40111789 90
M57958.1 - 529 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003073.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plaa

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
positive regulation of catalytic activity
protein binding
phospholipase A2 activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400024593.mm.1 NM_172695.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024593.mm.1 NM_172695.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400024593.mm.1 TC0400003073.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0