This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01256464_m1
See other RPE GE Assays ›
Gene Symbol:
Rpe
Gene Name:
ribulose-5-phosphate-3-epimerase
Gene Aliases:
2810429B02Rik, 5730518J08Rik
Chromosome Location:
Chr.1: 66700797 - 66719805 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01256464_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025683.2 NP_079959.2 2 - 3 219 59
GenBank mRNA AK007735.1 - 2 - 3 225 59
AK017774.1 - 2 - 3 220 59
AK029088.1 - 2 - 3 218 59
AK088152.1 - 2 - 3 220 59
AK150008.1 - 2 - 3 257 59
AK185217.1 - 2 - 3 213 59
BC019126.1 - 2 - 3 262 5008075 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000561.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpe

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
pentose-phosphate shunt
metabolic process
pentose-phosphate shunt, non-oxidative branch
pentose catabolic process
cellular carbohydrate metabolic process
catalytic activity
ribulose-phosphate 3-epimerase activity
isomerase activity
racemase and epimerase activity, acting on carbohydrates and derivatives
carbohydrate binding
identical protein binding
protein homodimerization activity
metal ion binding
monosaccharide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100004605.mm.1 NM_025683.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004605.mm.1 TC0100000561.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0