This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257218_mH
See other CEACAM3 GE Assays ›
Gene Symbol:
Ceacam3
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 3
Gene Aliases:
EG384557, Psg24, cea12
Chromosome Location:
Chr.7: 17150195 - 17163754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257218_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ceacam3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_054059.1 NP_473400.1 2 - 3 402 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000251.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700001870.mm.1 TC0700000251.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700001870.mm.1 NM_054059.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001870.mm.1 NM_054059.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001869.mm.1 TC0700000251.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700001869.mm.1 NM_054059.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001869.mm.1 NM_054059.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001867.mm.1 TC0700000251.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700001867.mm.1 NM_054059.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001867.mm.1 NM_054059.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0