This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257338_g1
See other SV2A GE Assays ›
Gene Symbol:
Sv2a
Gene Name:
synaptic vesicle glycoprotein 2 a
Gene Aliases:
AI746429, mKIAA0736
Chromosome Location:
Chr.3: 96181227 - 96195183 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257338_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sv2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022030.3 NP_071313.1 4 - 5 1384 78
GenBank mRNA AK028318.1 - 4 - 5 1329 78
AK080502.1 - 4 - 5 1337 78
AK080525.1 - 4 - 5 1426 78
AK122360.1 - 4 - 5 1420 78
AK144166.1 - 4 - 5 1407 78
BC046587.1 - 4 - 5 1384 6493509 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK158936.1 - Chr. - - [-] on Build 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000928.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sv2a

Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
cellular calcium ion homeostasis
chemical synaptic transmission
transmembrane transport
receptor activity
transporter activity
protein kinase binding
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300007430.mm.1 NM_022030.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300007430.mm.1 NM_022030.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300007430.mm.1 TC0300000928.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0